Learn More
Thermo Scientific™ Immuno Breakable Modules in White and Black
Keep wells in place with these breakable modules in white and black, ensuring fluorescence and luminescence assays are easy to handle.
£235.70 - £489.00
Product Code | Brand | Surface Treatment | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Color | Price | Quantity & Availability | ||||
10063743
|
Thermo Scientific™
446471 |
MaxiSorp | Black |
£471.00
Pack of 60 |
In Stock
Log in to see stock. |
||||
10521834
|
Thermo Scientific™
446473 |
PolySorp | Black |
£387.00
Pack of 60 |
In Stock
Log in to see stock. |
||||
10435205
|
Thermo Scientific™
463200 |
PolySorp | White |
£489.00
Pack of 60 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
10436473
|
Thermo Scientific™
463201 |
MaxiSorp | White |
£486.00
Pack of 60 |
In Stock
Log in to see stock. |
||||
10089174
|
Thermo Scientific™
95029800 |
Universal Binding | Black |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Keep wells securely in place with Thermo Scientific™ Breakable Modules in White and Black, ensuring fluorescence and luminescence assays are easy to handle.
Features:- High-quality polystyrene wells
- Choice of two passive surfaces to enable assay optimization
- C8 Strips provide flexiblity with sample size and optimal washing performance
- Easy to handle; single well held as easily as strips
- Compatible with common instrumentation
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays.
White:
- Opaque and reflective surface
- Low cross talk and minimal background
Black:
- Opaque and light-absorbing surface
- Minimum cross talk and back-scattered light
Note:
Availability may vary by country.
Specifications
Strip Plate | |
Polystyrene | |
None |