missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Product Code. 10673832
Click to view available options
Surface Treatment:
Enhanced Binding
Hydrophilic
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Volume (Metric) Working:
250 μL
330 μL
350 μL
Well Shape:
Curved Edges, Flat Bottom (C-well)
Round
Round, C Bottom
Round, Flat Bottom (F-well)
Well Design:
C12
C8
F12
F16 (2x8 wells)
F8
U16
U8
Unit Size:
Pack of 100
Pack of 120
Pack of 320
Pack of 50
Pack of 60
Pack of 640
This item is not returnable. View return policy

Product Code. 10673832

Brand: Thermo Scientific™ 469078

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat-bottom, C-bottom (radius edge), U-shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Certifications

Products specified in chart are certified for: Adsorption of IgG; CV of <5% between wells; all results +/-10% from mean; and background of all wells +/-0.005 absorbance units from mean.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Binding Property Hydrophobic
Certified No
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Microplate Strip
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 350 μL
Surface Treatment PolySorp
Well Shape Round, Flat Bottom (F-well)
No. per Case 60
No. per Pack 10
Plate Blocking None
Quantity 60/Cs.
Target Molecule Lipids
Well Design F8
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.