Learn More
Thermo Scientific™ Immuno Standard Modules White and Black, Immulon, F12, Microlite 2, 380μL
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo ScientificTM Immuno Standard Modules White and Black.
Brand: Thermo Scientific™ 7567
175.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 04
Additional Details : Weight : 1.80000kg
Description
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:- Available in eight-, 12- or 16-well strips
- Framed or unframed for choice of optimal assay
- Flat-bottom or C-shaped wells
- Choice of three surface types for optimal binding
- White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
- Black format to reduce back scatter light which may be encountered in epifluorescence
- Transparent or clear format for use in Time Resolved Fluorescence applications only
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
- Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
Specifications
Breakable Strips | |
No | |
Chemiluminescent | |
Adsorption | |
Microplate Strip | |
380 μL | |
330 μL | |
Luminescence | |
320 | |
320/Cs. |
Slightly Hydrophilic | |
Standard | |
320 Pk., 320 Cases | |
White | |
Polystyrene | |
96 | |
Round, Flat Bottom (F-well) | |
320 | |
None | |
Antibodies (Glycoproteins) and Proteins |