missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Standard Modules White and Black
Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
£267.00 - £1060.00
Specifications
Product Type | Microplate Strip |
---|---|
Material | Polystyrene |
No. of Wells | 96 |
Plate Blocking | None |
Product Code | Brand | Color | Volume (Metric) Well | Surface Treatment | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) Well | Surface Treatment | Price | Quantity & Availability | |||
10376231
|
Thermo Scientific™
437591 |
White | 350 μL | MaxiSorp |
£440.00
Pack of 60 |
In Stock
Log in to see stock. |
|||
10040591
|
Thermo Scientific™
437702 |
White | 350 μL | PolySorp |
£1060.00
Pack of 60 |
In Stock
Log in to see stock. |
|||
10080611
|
Thermo Scientific™
437915 |
Clear | 350 μL | MaxiSorp |
£374.00
Pack of 60 |
In Stock
Log in to see stock. |
|||
10759081
|
Thermo Scientific™
475515 |
Black | 400 μL | MaxiSorp |
£315.00
Pack of 60 |
In Stock
Log in to see stock. |
|||
10060591
|
Thermo Scientific™
475523 |
Black | 400 μL | PolySorp |
£306.00
Pack of 60 |
In Stock
Log in to see stock. |
|||
10634247
|
Thermo Scientific™
7566 |
White | 380 μL | Hydrophobic |
£267.00
Pack of 320 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
Description
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
- Available in 8-, 12- or 16-well strips
- Framed or unframed for choice of optimal assay
- Flat-bottom or C-shaped wells
- Choice of three surface types for optimal binding
- White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
- Black format to reduce backscatter light, which may be encountered in epifluorescence
- Transparent or clear format for use in time-resolved fluorescence applications only
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
- Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
Specifications
Microplate Strip | |
Polystyrene | |
96 | |
None |