missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Product Code. 10104421
Click to view available options
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
11 mm
12 mm
15 mm
Product Type:
Immuno Star Tubes and Stopper
Immuno Tubes and Stopper
Stopper
For Use With (Equipment):
11 mm Nunc-Immuno Tubes
12 mm Nunc-Immuno Tubes
15 mm Nunc-Immuno Tubes
Immuno Tube
Unit Size:
Pack of 1200
Pack of 1800
Pack of 2400
Pack of 3000
Pack of 3600
This item is not returnable. View return policy

Product Code. 10104421

Brand: Thermo Scientific™ 470319

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.

Features:
  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment

Choice of Surface:

  • Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
  • Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
  • Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.

Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):

  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions

Solid Phase Immunoassay Tubes:

  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • Certified binding homogeneity
  • High–quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times

Recommended for:

  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
TRUSTED_SUSTAINABILITY

Specifications

Material Polystyrene
Product Type Immuno Star Tubes and Stopper
Surface MaxiSorp
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
Certifications/Compliance MaxiSorp tubes are certified for IgG binding (tube to tube % CV less than 5%)
No. per Pack 100
No. per Case 3000
Volume (Metric) 5 mL
For Use With (Equipment) Immuno Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.