missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Volume (Metric) | Material | Diameter (Metric) Outer | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Material | Diameter (Metric) Outer | Product Type | Price | Quantity & Availability | |||||
10316141
|
Thermo Scientific™
341866 |
Not Applicable | Polyethylene | 11 mm | Stopper |
£199.00
Pack of 3600 |
In Stock
Log in to see stock. |
|||||
10172831
|
Thermo Scientific™
343036 |
Not Applicable | Polyethylene | 15 mm | Stopper |
POA
Pack of 2400 |
N/A | |||||
10736662
|
Thermo Scientific™
348801 |
Not Applicable | Polyethylene | 12 mm | Stopper |
£159.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||||
10254071
|
Thermo Scientific™
443990 |
5 mL | Polyethylene | 12 mm | Immuno Tubes and Stopper |
£319.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||||
10419962
![]() |
Thermo Scientific™
444202 |
5 mL | Polystyrene | 12 mm | Immuno Tubes and Stopper |
£258.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||||
10596171
|
Thermo Scientific™
444474 |
4 mL | Polystyrene | 11 mm | Immuno Tubes and Stopper | N/A | ||||||
10643452
|
Thermo Scientific™
466982 |
4 mL | Polyethylene | 11 mm | Immuno Tubes and Stopper | N/A | ||||||
10419002
|
Thermo Scientific™
468608 |
12 mL | Polyethylene | 15 mm | Immuno Tubes and Stopper |
£211.00
Pack of 1200 |
In Stock
Log in to see stock. |
|||||
10104421
|
Thermo Scientific™
470319 |
5 mL | Polystyrene | 12 mm | Immuno Star Tubes and Stopper |
£298.00
Pack of 3000 |
In Stock
Log in to see stock. |
|||||
Description
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:- Offered in choice of three surfaces for optimal binding
- Three tube sizes with stoppers available (ordered separately)
- Fits standard equipment
Choice of Surface:
- Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
- Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
- Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
- MiniSorp surface
- Molded from special polyethylene formulation
- For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
- PolySorp and MaxiSorp surfaces available for choice of optimal binding
- For solid phase immuno techniques such as IRMA, ELISA and ILMA
- High–quality polystyrene for optimal quality
- StarTube design gives higher sensitivity and faster assay times
- Enhances assay sensitivity and reduces incubation times
Recommended for:
- ELISA
- Solid Phase RIA
- Liquid Phase RIA
- Sample storage
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Immuno Tubes and Stoppers