missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ImmunoCult™ Human CD3/CD28 T Cell Activator

Product Code. p-7217507
Encompass
Click to view available options
Quantity:
2 mL
5 x 2 mL
Unit Size:
2mL
Pack of 5
This item is not returnable. View return policy

Product Code. 16425234

Brand: STEMCELL Technologies 10991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human T cell activation and expansion reagent

  • Achieve robust activation and expansion of T cells in the absence of magnetic beads, feeder cells, or antigens
  • This product's gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded, for example in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981)
  • Tetrameric antibody complexes bind to and cross-link CD3 and CD28 cell surface ligands, thereby providing the required signals for T cell activation
  • ImmunoCult™ Human CD3/CD28 T Cell Activator can be used on the Seahorse XF Analyzer to measure T cell activation response and is also available as part of the Agilent Seahorse XF Hu T Cell Activation Assay Kit
  • This product is designed for cell therapy research applications, but may be qualified for use as an ancillary material (AM) following the framework outlined in USP<1043>
  • STEMCELL Technologies can work with you to qualify this reagent as an AM under an approved investigational new drug (IND) or clinical trial application (CTA)

Specifications

Cell Type T Cells, T Cells, CD4+, T Cells, CD8+
For Use With (Application) Activation, Cell Culture, Expansion
Quantity 5 x 2 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.