Learn More
Ohaus™ Impact Printer
Combines high performance and multiple functions to offer OHAUS quality and durability in a portable printer. Ohaus™ Impact Printers offer advanced functionality in an easy-to-use and set up printer at an exceptional value.
Brand: Ohaus™ 30064202
614.08 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Ideal for use in conjunction with OHAUS balances and scales in laboratory, industrial, and educational applications to ensure accurate documentation of weighing results.- AC adapter included
- RS233 communication
- ABS Housing
- Status indicator, 40 characters per line, compatible with continuous label paper, perform zero and tare operations directly from the printer
Specifications
180 x 106 x 88 mm | |
88 mm | |
Impact Printer |
1.6 mm | |
1 Each |