missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Impeller Sleeve for Open-Top Mixing, for 200 L HyPerforma™ Single-Use Mixer BPC

Support impeller-based mixing in bioproduction using these bioprocess containers.

Brand:  Thermo Scientific™ SH30749.08

Product Code. 30040284

  • £1790.00 / Pack of 8

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Single Use Mixer (S.U.M.) BPC options include:
  • Open top liner for media/buffer hydration
  • Closed BPC with powder port for contained media/buffer hydration
  • Closed BPC with powder port and monitoring probe capability
  • Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
  • Closed BPC for liquid-liquid mixing with monitoring probe capability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HyPerforma 50 L Single-Use Mixer, DS 300 Single-Use Mixer
Sterile
HyPerforma™
Impeller Sleeve
200 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers