missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ In-flow Safety Cans
For use in storing reagents in approved containers when drawing solvents into HPLC machines
Brand: Justrite™ 12164
This item is not returnable.
View return policy
Description
- Made of high density polyethylene
- Equipped with a required self-closing fill/pour spout with stainless steel flame arrester
- Two quick-disconnect fittings: one polypropylene for venting, and one stainless steel to handle the solvent in-flow
- Stainless steel disconnect includes 1/8 in. OD (1/16 in.ID) and 1/16 in. OD (1/32 in. ID) compression fittings
Specifications
FM, NFPA, OSHA | |
0.5 gal. | |
White | |
Self Closing Fill/Pour Spout | |
5.25 in. | |
9.5 in. | |
1.45 in. | |
3.6 cm |
Safety Can | |
2 L | |
Polyethylene | |
Oval | |
13.3 cm | |
24.1 cm | |
7.75 in. | |
19.6 mm |