missing translation for 'onlineSavingsMsg'
Learn More

Indene, 90%, tech., stabilized, Thermo Scientific Chemicals

Product Code. 10245210
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-13-6, 98-29-3
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00003777
InChI Key YBYIRNPNPLQARY-UHFFFAOYSA-N
Synonym indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy
PubChem CID 7219
ChEBI CHEBI:41921
IUPAC Name 1H-indene
SMILES C1C=CC2=CC=CC=C21

Specifications

Melting Point -5°C to 3°C
Color White to Yellow
Density 0.9900g/mL
Boiling Point 181.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5670 to 1.5720
Quantity 5 g
Beilstein 05, 515
Merck Index 15, 4978
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents,soluble in acetone,miscible with ethanol and ethyl ether
Stabilizer 5 to 10ppm tert-butylcatechol
Formula Weight 116.16
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Indene, tech., 90%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'