Learn More
Ismatec™ Independent Channel Control Peristaltic Pump Assembly
Expand your pumping capabilities with a single compact unit. Ismatec™ Independent Channel Control Peristaltic Pump Assembly delivers continuous pumping or precision dispensing up to 35mL/min with the capability of bidirectional flow in each channel.
Brand: Ismatec™
Code : NEW
Additional Details : Weight : 2.70000kg
Specifications
4 | |
IP 30 | |
14.5 psi | |
8 | |
2.7 kg | |
50/60 Hz |
3 stop color coded | |
35 mL/min. | |
0.1 to 100 rpm | |
182 x 127 x 145 mm | |
30 w |