missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo carmine

Artikelnummer. 11428716
Click to view available options
Quantity:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Useful in the staining of Negri bodiesIndigo carmine is used as a food colorant, pH indicator and dissolved ozone indicator. It is used to detect amino fluid leaks, highlight portions of the urinary tract, tissue stains and tool for the determination of hypochlorite. As a dye in renal function, it is used in testing and detection of nitrates and chlorates, and in the testing of milk.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful in the staining of Negri bodiesIndigo carmine is used as a food colorant, pH indicator and dissolved ozone indicator. It is used to detect amino fluid leaks, highlight portions of the urinary tract, tissue stains and tool for the determination of hypochlorite. As a dye in renal function, it is used in testing and detection of nitrates and chlorates, and in the testing of milk.

Solubility
Soluble in ethylene glycol monomethyl ether and ethanol. Slightly soluble in water and alcohol. Insoluble in benzene, chloroform and organic solvents.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym indigo carmine, indigocarmine, acid blue 74, indigocarmin, indigotindisulfonate sodium, c.i. acid blue 74, food blue no. 2, amacid brilliant blue, c.i. food blue 1, fd&c blue no. 2
PubChem CID 5284351
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Spezifikation

Melting Point >300°C
Odor Odorless
Quantity 25 g
Beilstein 4103904
Sensitivity Light sensitive
Merck Index 14,4944
Solubility Information Soluble in ethylene glycol monomethyl ether and ethanol. Slightly soluble in water and alcohol. Insoluble in benzene,chloroform and organic solvents.
IUPAC Name disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
Formula Weight 466.36
Chemical Name or Material Indigo carmine
compliance-icons
Signalwort
  • Warning
Gefahrenkategorie
  • Skin sensitiser Category 1
Gefahrenhinweis
  • H317-May cause an allergic skin reaction.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'