missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo Carmine

Catalog Number 10381731
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
860-22-0
C16H8N2Na2O8S2
466.35
MFCD00005723
KHLVKKOJDHCJMG-QDBORUFSSA-L
Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt
5284351
disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
This item is not returnable. View return policy
860-22-0
C16H8N2Na2O8S2
466.35
MFCD00005723
KHLVKKOJDHCJMG-QDBORUFSSA-L
Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt
5284351
disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Absorption Ratio 0.98 to 1.07
Dye Content 80% min.
Beilstein 25, 304
Merck Index 15, 4982
Solubility Information Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents
Infrared Spectrum Authentic
Lambda Max 608 to 612nm
Formula Weight 466.34
Grade Pure
Packaging Glass bottle
Color Blue to Purple
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Indigo carmine, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Indigo Carmine
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'