missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo, synthetic

Product Code. 10206940
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 482-89-3
Molecular Formula C16H10N2O2
Molecular Weight (g/mol) 262.268
MDL Number MFCD00005722
InChI Key COHYTHOBJLSHDF-BUHFOSPRSA-N
Synonym C.I. 73000, Vat Blue 1
PubChem CID 5318432
IUPAC Name (2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one
SMILES C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2

Specifications

Beilstein 24, 417
Merck Index 15, 4981
Infrared Spectrum Authentic
Formula Weight 262.26
Packaging Glass bottle
UV Spectrometry Complies
Color Blue to Violet
Quantity 25 g
Physical Form Powder
Chemical Name or Material Indigo, synthetic
compliance-icons
Product Identifier
  • Indigo
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.