Learn More
THERMOFISHER SCIENTIFIC AUSTRALIA PTY LTD Indiko Wash Solution
Indiko Wash Solution 6x100mL
Brand: THERMOFISHER SCIENTIFIC AUSTRALIA PTY LTD 980929
Code : NEW
Additional Details : CAS Number : 7681-52-9 Weight : 0.99000kg Transport : UN number : 1791 Chem class : 8 Pack group : III
Description
Indiko Wash Solution 6x100mL
- Indiko Wash Solution
- 6x100 mL
Specifications
Wash Solution | |
Wash solution | |
6 x 100 mL |
Safety and Handling
- Washing solution 4.5%
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sodium hypochlorite