missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium, 99.999%, (trace metal basis), powder
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Specifications
| Color | Gray |
| Melting Point | 156.0°C |
| Boiling Point | 2000.0°C |
| Density | 7.3000g/mL |
| Quantity | 25 g |
| Packaging | Glass bottle |
| Assay Percent Range | 100.00% |
| Form | Powder |
| Linear Formula | In |
| Fieser | 06,293; 14,181; 16,181; 17,152 |
| Show More |
Safety and Handling
Product Identifier
- Indium, powder
- Warning
- Acute toxicity Category 4
- Flammable solid Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction