missing translation for 'onlineSavingsMsg'
Learn More

Indium antimonide, 99.999% (metals basis)

Product Code. 11322588
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11322588

Brand: Thermo Scientific Alfa Aesar 014626.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1312-41-0 | InSb | 236.578 g/mol

Indium antimonide finds use in infrared detectors, including FLIR systems, thermal imaging cameras, infrared astronomy and in infrared homing missile guidance systems, in fast transistors. It is used in thermal image detectors using photo-electromagnetic detectors or photodiodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-41-0
Molecular Formula InSb
Molecular Weight (g/mol) 236.578
MDL Number MFCD00016146
InChI Key WPYVAWXEWQSOGY-UHFFFAOYSA-N
PubChem CID 3468413
IUPAC Name indiganylidynestibane
SMILES [In]#[Sb]

Specifications

Melting Point 535°C
Physical Form Pieces
Quantity 10 g
UN Number UN1549
Merck Index 14,4948
Solubility Information Insoluble in water.
Formula Weight 236.57
Percent Purity 99.999%
Odor Odorless
Density 5.75 g/mL at m.p.
Assay (metals basis)
Chemical Name or Material Indium antimonide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.