missing translation for 'onlineSavingsMsg'
Learn More
Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals Product Code.: 11341179

Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals

Product Code. 11341179
100 x 200 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
100 x 200 mm
This item is not returnable. View return policy

Product Code. 11341179

Brand: Thermo Scientific Alfa Aesar 011384.GK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronicShow MoreShow Less
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Quantity 100 x 200 mm
Percent Purity 99.9975%
Grade Puratronic™
Odor Odorless
Weight ≈2.3g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Indium foil, 0.5mm (0.02 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM21L014 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number011384.FI,011384.FY,011384.GK,011384.FF
Lot NumberM21L015 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number011384.FI,011384.FY,011384.GK,011384.FF
Lot NumberU05K038 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number011384.FI,011384.FY,011384.GK,011384.FF
Lot NumberW19K050 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number011384.FI,011384.FY,011384.GK,011384.FF
Lot NumberU05K037 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number011384.FI,011384.FY,011384.GK,011384.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.