Learn More
Indium(I) chloride, anhydrous, 99.995% (metals basis), Thermo Scientific Chemicals
Description
Indium chloride is a lewis acid catalyst in organic reactions such as Friedel-Crafts acylations and Diels-Alder reactions and it is used in high-performing solar cells such as optics and microelectronics. It applications finds in semiconductor technology, producing phosphor,low pressure sodium lamp and produce high purity chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon

Specifications
Specifications
Boiling Point | 608°C |
Melting Point | ∼225°C |
Physical Form | Crystalline |
Quantity | 25 g |
UN Number | UN3260 |
Beilstein | EM>13, |
Solubility Information | Soluble in water and THF. |
Formula Weight | 150.27 |
Percent Purity | 99.995% |
Odor | Chlorine-like |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W16K005 | Certificate of Analysis | 30/09/2024 | 040033.02,040033.14,040033.06 |
M16K006 | Certificate of Analysis | 23/01/2024 | 040033.02,040033.14,040033.06 |
Z19I046 | Certificate of Analysis | 13/01/2023 | 040033.02,040033.14,040033.06 |
P21I080 | Certificate of Analysis | 12/04/2022 | 040033.02,040033.14,040033.06 |
B14R052 | Certificate of Analysis | 08/12/2021 | 040033.02,040033.14,040033.06 |
Safety and Handling

- Indium chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Indium chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.