missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) chloride, Puratronic™, 99.999% (metals basis)

Product Code. 11320979
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11320979

Brand: Thermo Scientific Alfa Aesar 010707.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13465-10-6 | ClIn | 150.27 g/mol

Indium(I) chloride is used to prepare cage, dinuclear complexes (In-cyclodiphophazanes) which possess interesting metal-metal interactions, Semiconductor doping, phosphors-compound semiconductors, ITO thin-film, Basic raw material for compounding organic indium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-10-6
Molecular Formula ClIn
Molecular Weight (g/mol) 150.27
MDL Number MFCD00148875
InChI Key APHGZSBLRQFRCA-UHFFFAOYSA-M
IUPAC Name λ¹-indiganyliumylidene chloride
SMILES [Cl-].[In+]

Specifications

Boiling Point 608°C
Melting Point ∼225°C
Physical Form Crystalline Lump
Quantity 25 g
UN Number UN3260
Solubility Information Decomposes in water.
Formula Weight 150.27
Percent Purity 99.999%
Grade Puratronic
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 4.19 g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.