Learn More
Indium(I) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Indium(I) iodide can be used to prepare indium cluster and chain compounds. It is also used as a reagent to propagate organic reactions viz. cyclization and cleavage of diselenide or disulfide. It plays an important role in preparing indium containing complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon

Specifications
Specifications
Melting Point | 351°C |
Physical Form | Beads |
Quantity | 5 g |
Solubility Information | Slightly soluble in some organic solvents,pyridine or m-xylene mixture. |
Formula Weight | 241.72 |
Percent Purity | 99.998% |
Packaging | Ampule |
Sensitivity | Light and moisture sensitive |
Assay | (metals basis) |
Chemical Name or Material | Indium(I) iodide, Ultra dry |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z27K049 | Certificate of Analysis | 23/01/2025 | 035700.03,035700.06 |
Z27K012 | Certificate of Analysis | 23/01/2025 | 035700.03,035700.06 |
Y12K022 | Certificate of Analysis | 16/12/2024 | 035700.03,035700.06 |
U07K039 | Certificate of Analysis | 07/10/2024 | 035700.03,035700.06 |
U07K018 | Certificate of Analysis | 07/10/2024 | 035700.03,035700.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.