missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) acetate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11379877
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Indium(III) acetate is used as a catalyst in intermolecular radical addition of organic iodides to electron deficient alkenes. It is also used in the reduction of 1-bromo-3-phenylpropane to propylbenzene. Further, it is used to prepare the indium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25114-58-3
Molecular Formula C2H4InO2
Molecular Weight (g/mol) 174.87
MDL Number MFCD00054347
InChI Key LGHOYKSQIQISBI-UHFFFAOYSA-N
Synonym indium acetate, acetic acid, indium 3+ salt
PubChem CID 24191761
IUPAC Name acetic acid;indium
SMILES CC(=O)O.[In]

Specifications

Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (Metals Basis)
Solubility Information Soluble in water,mineral acid and acetic acid.
Formula Weight 291.96
Sensitivity Hygroscopic
Chemical Name or Material Indium(III) Acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'