Learn More
Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
£65.83 - £217.08
Chemical Identifiers
CAS | 25114-58-3 |
---|---|
Molecular Formula | C6H9InO6 |
Molecular Weight (g/mol) | 291.95 |
InChI Key | LGHOYKSQIQISBI-UHFFFAOYSA-N |
Synonym | indium acetate, acetic acid, indium 3+ salt |
PubChem CID | 24191761 |
IUPAC Name | acetic acid;indium |
SMILES | CC(=O)O.[In] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11975921
|
Thermo Scientific Acros
436980100 |
10 g | Glass bottle |
£65.83
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11985921
|
Thermo Scientific Acros
436980500 |
50 g | Glass bottle |
£217.08
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
25114-58-3 | |
291.95 | |
indium acetate, acetic acid, indium 3+ salt | |
acetic acid;indium |
C6H9InO6 | |
LGHOYKSQIQISBI-UHFFFAOYSA-N | |
24191761 | |
CC(=O)O.[In] |
Specifications
270.0°C | |
White | |
35.3 to 43.3% (In) (Complexometry) | |
indium acetate, acetic acid, indium 3+ salt | |
LGHOYKSQIQISBI-UHFFFAOYSA-N | |
acetic acid;indium | |
24191761 | |
99.99% | |
Indium(III) acetate |
25114-58-3 | |
Crystalline Powder | |
C6H9InO6 | |
Solubility in water: soluble | |
CC(=O)O.[In] | |
291.95 | |
291.95 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
- Indium(III) acetate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only