missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) bromide, anhydrous, 99.99% (metals basis)

Product Code. p-7050967
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11339267

Brand: Thermo Scientific Alfa Aesar 040126.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13465-09-3

Indium(III) bromide is used in organic synthesis as a water tolerant Lewis acid.It efficiently catalyzes the three-component coupling of β-keto esters, aldehydes and urea (or thiourea) to afford the corresponding dihydropyrimidinones. It is used as a catalyst to produce dithioacetals when unactivated alkynes react with thiols and fields such as optics and microelectronics that utilize semiconductor technology have wide uses for indium in high-performing solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-09-3
MDL Number MFCD00011057

Specifications

Melting Point ∼436°C
Color White to Yellow
Physical Form Crystalline Powder and/or Chunks
Quantity 10 g
Linear Formula InBr3
Solubility Information Soluble in water.
Formula Weight 354.55
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.74 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.