missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11364967

Indium(III) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11364967
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11364967

Brand: Thermo Scientific Alfa Aesar 041977.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indium(III) chloride is used in organic reactions such as Friedel-crafts acylation and Diels-Alder reaction as a Lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-82-8
Molecular Formula InCl3
MDL Number MFCD00011058

Specifications

Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water and tetrhydrofuran. Sparingly soluble in alcohol. Insoluble in ether.
Formula Weight 221.18
Sensitivity Hygroscopic
Density 3.46
Chemical Name or Material Indium(III) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU26K015 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number041977.18,041977.09,041977.30
Lot NumberM25K002 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number041977.18,041977.09,041977.30
Lot NumberW06J003 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number041977.18,041977.09,041977.30
Lot NumberR16I032 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog Number041977.18,041977.09,041977.30
Lot NumberT08I033 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog Number041977.18,041977.09,041977.30
compliance-icons compliance-icons
Product Identifier
  • Indium(III) chloride anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Indium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.