missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11321928

Indium(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11321928
250 g
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11321928

Brand: Thermo Scientific Alfa Aesar 011856.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-82-8
MDL Number MFCD00011058

Specifications

Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Crystalline
Quantity 250 g
UN Number UN3260
Merck Index 14,4958
Solubility Information It is very soluble in water and THF. It is sparingly soluble in alcohol and insoluble in ether.
Formula Weight 221.18
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Density 3.460 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) chloride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY08K029 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number011856.30,011856.03,011856.09,011856.18
Lot NumberX02K031 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number011856.30,011856.03,011856.09,011856.18
Lot NumberS24K053 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number011856.30,011856.03,011856.09,011856.18
Lot NumberT24K024 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number011856.30,011856.03,011856.09,011856.18
Lot NumberQ15K025 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011856.30,011856.03,011856.09,011856.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.