missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11331928
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-82-8
MDL Number MFCD00011058

Specifications

Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Merck Index 14,4958
Solubility Information It is very soluble in water and THF. It is sparingly soluble in alcohol and insoluble in ether.
Formula Weight 221.18
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Density 3.460 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'