missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog Number 11438490
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10025-82-8
InCl3
MFCD00011058
This item is not returnable. View return policy
10025-82-8
InCl3
MFCD00011058

Indium(III) chloride is used as catalyst for aqueous organic reactions including C-C bond formation, aldol reactions, and reductions and for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Reacts with water.
Formula Weight 221.18
Sensitivity Hygroscopic
Density 3.46
Chemical Name or Material Indium(III) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'