missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) fluoride, anhydrous, 96%, Thermo Scientific Chemicals

£54.17 - £210.05

Chemical Identifiers

CAS 7783-52-0
MDL Number MFCD00016149
Synonym indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389247
View Documents
Thermo Scientific Alfa Aesar
040118.04
2 g
£54.17
2g
Please to purchase this item. Need a web account? Register with us today!
11379247
View Documents
Thermo Scientific Alfa Aesar
040118.09
10 g
£210.05
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium(III) fluoride is employed in the synthesis of non-oxide glasses. It promotes chemoselective addition of TMSCN to aldehydes forming the respective cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-52-0
MFCD00016149
indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride

Specifications

>1200°C
7783-52-0
Powder
14,4959
Slightly soluble in water (0;04g/100mL); Freely soluble in dilute acids
96%
Moisture sensitive
Indium(III) fluoride, Anhydrous
∼1170°C
White
MFCD00016149
indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride
171.82
Odorless
4.39 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H332-H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 232-005-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Indium(III) fluoride, anhydrous

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Indium(III) fluoride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only