Learn More
Indium(III) iodide, 99.998%, (trace metal basis), Thermo Scientific Chemicals
£46.93 - £320.60
Chemical Identifiers
CAS | 13510-35-5 |
---|---|
Molecular Formula | InI3 |
Molecular Weight (g/mol) | 495.53 |
MDL Number | MFCD00016151 |
InChI Key | RMUKCGUDVKEQPL-UHFFFAOYSA-K |
Synonym | indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis |
PubChem CID | 83539 |
IUPAC Name | triiodoindigane |
SMILES | [In](I)(I)I |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10553421
|
Thermo Scientific Acros
316010010 |
1 g | Glass bottle |
£46.93
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10202992
|
Thermo Scientific Acros
316010050 |
5 g | Glass bottle |
£149.75
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10711365
|
Thermo Scientific Acros
316010250 |
25 g | Glass bottle |
£320.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
13510-35-5 | |
495.53 | |
RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
83539 | |
[In](I)(I)I |
InI3 | |
MFCD00016151 | |
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
triiodoindigane |
Specifications
210.0°C | |
Brown-Yellow to Yellow | |
22.7 to 23.5% (In) (Complexometry) | |
InI3 | |
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
triiodoindigane | |
83539 | |
99.998% | |
Indium(III) iodide |
13510-35-5 | |
Crystals or Powder | |
InI3 | |
MFCD00016151 | |
Solubility in water: soluble | |
[In](I)(I)I | |
495.53 | |
495.53 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 236-839-6
RUO – Research Use Only