missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) nitrate hydrate, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11359317

Indium(III) nitrate hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11359317
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11359317

Brand: Thermo Scientific Alfa Aesar 040185.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207398-97-8
Molecular Formula InN3O9
Molecular Weight (g/mol) 300.83
MDL Number MFCD00149742
InChI Key LKRFCKCBYVZXTC-UHFFFAOYSA-N
Synonym indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2oShow MoreShow Less
PubChem CID 3084148
SMILES [In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 100°C-H2O
Color White
Physical Form Lump/Powder
Quantity 50 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,methanol and ethanol.
IUPAC Name indium(3+) trinitrate
Formula Weight 300.83 (Anhydrous)
Sensitivity Hygroscopic
Appearance White to off-white lump/powder
Chemical Name or Material Indium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY27K008 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog Number040185.18,040185.09
Lot NumberY08K043 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number040185.18,040185.09
Lot NumberW26K035 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number040185.18,040185.09
Lot NumberT26K041 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number040185.18,040185.09
Lot NumberZ11J044 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number040185.18,040185.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) nitrate hydrate, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.