missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, 99.9% (metals basis)

Product Code. 11357427
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11357427

Brand: Thermo Scientific Alfa Aesar 012581.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

Indium(III) oxide is widely utilized as a n-type semiconductor, which is used in integrated circuits as a resistive element. It finds applications in batteries and as a part of some stain formulations. It is used as thin film coatings in optical, antistatic and infrared reflectors. It is doped with tin oxide and used as transparent conductive coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850° volat.
Color Yellow
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.994% (Metal basis)
Solubility Information Insoluble in water and soluble in acids.
Formula Weight 277.64
Odor Odorless
Appearance Yellow powder
Density 7.18
Chemical Name or Material Indium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.