missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) phosphide, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11323358

Indium(III) phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11323358
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11323358

Brand: Thermo Scientific Alfa Aesar 036283.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The water soluble InP/ZnS core/shell QDs are a safer alternative to CdSe/ZnS QDs for biological applications, by comparing their toxicity in vitro (cell culture) and in vivo.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22398-80-7
Molecular Formula InP
Molecular Weight (g/mol) 145.792
MDL Number MFCD00016153
InChI Key GPXJNWSHGFTCBW-UHFFFAOYSA-N
Synonym indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875Show MoreShow Less
PubChem CID 31170
IUPAC Name indiganylidynephosphane
SMILES P#[In]

Specifications

Melting Point 1070°C
Quantity 1 g
Merck Index 14,4953
Solubility Information Insoluble in water.
Formula Weight 145.79
Percent Purity 99.999%
Density 4.81 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) phosphide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF24P22 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036283.03,036283.06
Lot NumberG20R005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036283.03,036283.06
Lot NumberY13E006 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036283.03,036283.06
Lot NumberI19L38 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036283.03,036283.06
Lot NumberK14T038 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036283.03,036283.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) phosphide, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.