missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

£72.16 - £295.47

Chemical Identifiers

CAS 13464-82-9
Molecular Formula In2O12S3
Molecular Weight (g/mol) 517.80
MDL Number MFCD00011059
InChI Key XGCKLPDYTQRDTR-UHFFFAOYSA-H
Synonym indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
PubChem CID 26044
IUPAC Name indium(3+);trisulfate
SMILES [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11390979
View Documents
Thermo Scientific Alfa Aesar
010711.09
10 g
£72.16
10g
Please to purchase this item. Need a web account? Register with us today!
11380979
View Documents
Thermo Scientific Alfa Aesar
010711.18
50 g
£295.47
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium(III) sulfate is used Chemical synthesis, hardening agent in gold electroplating baths. It is also used to prepare other indium containing substances such as copper indium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13464-82-9
517.80
XGCKLPDYTQRDTR-UHFFFAOYSA-H
26044
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
In2O12S3
MFCD00011059
indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
indium(3+);trisulfate

Specifications

250°C (decomposition)
Lump/Powder
MFCD00011059
Soluble in water and ethanol. Insoluble in basic salts.
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
517.80
517.83 (Anhydrous)
Puratronic™
Indium(III) sulfate hydrate
13464-82-9
In2O12S3
indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
XGCKLPDYTQRDTR-UHFFFAOYSA-H
indium(3+);trisulfate
26044
99.999%
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 236-689-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only