missing translation for 'onlineSavingsMsg'
Learn More
Indium shot, 5mm (0.2in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11300789

Indium shot, 5mm (0.2in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11300789
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11300789

Brand: Thermo Scientific Alfa Aesar 010615.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronicShow MoreShow Less
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver-White
Appearance Silvery-white shot
Assay Percent Range 99.9999% (Metals basis)
Quantity 250 g
Odor Odorless
Form Shot
Chemical Name or Material Indium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS30I034 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number010615.18,010615.09,010615.30,010621.22
Lot NumberJ26X003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010615.18,010615.09,010615.30,010621.22
Lot NumberL14Q029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010615.18,010615.09,010615.30,010621.22
Lot NumberF09F17 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010615.18,010615.09,010615.30,010621.22
Lot NumberA19U050 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010615.18,010615.09,010615.30,010621.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium shot, 5mm (0.2in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.