missing translation for 'onlineSavingsMsg'
Learn More
Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11396567

Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11396567
1 m
Click to view available options
Quantity:
1 m
5 m
This item is not returnable. View return policy

Product Code. 11396567

Brand: Thermo Scientific Alfa Aesar 000470.G1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 5.74g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronicShow MoreShow Less
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver-White
Appearance Silvery-white wire
Density 7.31g/cm3 at 20°C
Assay Percent Range 99.999% (Metals basis)
Quantity 1 m
Odor Odorless
Form Wire
Chemical Name or Material Indium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY02H043 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000470.G1,000470.G5
Lot NumberB24P25 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000470.G1,000470.G5
Lot NumberP16H080 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000470.G1,000470.G5
Lot NumberX02F038 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000470.G1,000470.G5
Lot NumberL17Y024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000470.G1,000470.G5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.