Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Fire Fighting
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand:  Brady™ N008551
 View more versions of this product
Product Code. 15329265
  • £131.00 / 1 roll
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Fire Fighting - Foam
Silicone
Roll
Pipe Marker
Pipe and valve marking
30 Markers/Roll
-40°F to +284°F
0.0047 in.
33 m x 355 mm
33 m
355 mm
37 mm
FOAM 355X37RL-T1-LL-P04
Permanent Acrylic, Permanent, Pressure-sensitive
White on Red
Polyester
Foam
English
30 Pk.
-40°C to +140°C
0.119 mm
36.09 yd.
13.98 in.
1.46 in.
16.5 mm