missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Flammable/Non-Flammable Liquids/Oils
European Standard Pipemarkers reinforce safety and aid maintenance.
£102.00 - £154.00
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC |
---|---|
Coating | Silicone |
Color | White on Dark Brown |
Format | Roll |
Material | Polyester |
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
15399135
|
Brady™
N007921 |
Emulsion | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319145
|
Brady™
N007922 |
Emulsion | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339185
|
Brady™
N008116 |
Hydraulic oil | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329145
|
Brady™
N007923 |
Emulsion | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349185
|
Brady™
N008117 |
Hydraulic oil | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379175
|
Brady™
N008107 |
Lubricating oil | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359185
|
Brady™
N008118 |
Hydraulic oil | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389175
|
Brady™
N008108 |
Lubricating oil | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319175
|
Brady™
N008089 |
Oil | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399175
|
Brady™
N008109 |
Lubricating oil | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329175
|
Brady™
N008090 |
Oil | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339175
|
Brady™
N008091 |
Oil | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC | |
White on Dark Brown | |
Polyester | |
Pipe and valve marking | |
30 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
36.09 yd. |
Silicone | |
Roll | |
Pipe Marker | |
English | |
30 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m |