missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads, with Pictograms - Flammable/Non Flammable Liquids
European Standard Pipemarkers reinforce safety and aid maintenance.
£102.00 - £154.00
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC |
---|---|
Adhesive Type | Permanent Acrylic, Pressure-sensitive |
Coating | Silicone |
Color | White on Dark Brown |
Display | Warning |
Product Code | Brand | Display Text | Width (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Width (Metric) | Height (Metric) | Price | Quantity & Availability | |||
15399195
|
Brady™
N008278 |
White Spirit | 250 mm | 26 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309205
|
Brady™
N008279 |
White Spirit | 355 mm | 37 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399155
|
Brady™
N008060 |
Diesel Fuel | 250 mm | 26 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319205
|
Brady™
N008280 |
White Spirit | 450 mm | 52 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309165
|
Brady™
N008061 |
Diesel Fuel | 355 mm | 37 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319165
|
Brady™
N008062 |
Diesel Fuel | 450 mm | 52 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359165
|
Brady™
N008071 |
Diesel | 250 mm | 26 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369165
|
Brady™
N008072 |
Diesel | 355 mm | 37 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309155
|
Brady™
N007954 |
Fuel | 250 mm | 26 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379165
|
Brady™
N008073 |
Diesel | 450 mm | 52 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319155
|
Brady™
N007955 |
Fuel | 355 mm | 37 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329155
|
Brady™
N007956 |
Fuel | 450 mm | 52 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC | |
Silicone | |
Warning | |
Polyester | |
Pipe and valve marking | |
30 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
36.09 yd. |
Permanent Acrylic, Pressure-sensitive | |
White on Dark Brown | |
Roll | |
Pipe Marker | |
English | |
30 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m |