missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads, with Pictograms - Gas

European Standard Pipemarkers reinforce safety and aid maintenance.

Brand:  Brady™ N007439

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15359055

  • £154.00 / 1 roll
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC
Permanent Acrylic, Pressure-sensitive
Black on Ochre
Roll
Pipe Marker
Pipe and valve marking
30 Markers/Roll
-40°F to +284°F
0.0047 in.
33 m x 450 mm
33 m
450 mm
52 mm
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, with pictograms - Gas - CO2
Silicone
Warning
Polyester
CO2
English
30 Pk.
-40°C to +140°C
0.119 mm
36.09 yd.
17.72 in.
2.05 in.
22.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers