Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Water (A-H)
European Standard Pipemarkers reinforce safety and aid maintenance.
£102.00 - £154.00
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC |
---|---|
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Coating | Silicone |
Color | White on Green |
Format | Roll |
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
15359405
|
Brady™
N009145 |
Canal water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349405
|
Brady™
N009144 |
Canal water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369405
|
Brady™
N009146 |
Canal water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369455
|
Brady™
N009342 |
Chilled water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389455
|
Brady™
N009344 |
Chilled water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379455
|
Brady™
N009343 |
Chilled water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399335
|
Brady™
N008866 |
Chlorated water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389335
|
Brady™
N008865 |
Chlorated water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309345
|
Brady™
N008867 |
Chlorated water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379415
|
Brady™
N009180 |
Cold water return | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389415
|
Brady™
N009181 |
Cold water return | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309415
|
Brady™
N009162 |
Cold water supply | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399415
|
Brady™
N009182 |
Cold water return | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319415
|
Brady™
N009163 |
Cold water supply | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329415
|
Brady™
N009164 |
Cold water supply | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359295
|
Brady™
N008658 |
Condensate | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369295
|
Brady™
N008659 |
Condensate | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379295
|
Brady™
N008660 |
Condensate | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339365
|
Brady™
N009009 |
Cooling water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349365
|
Brady™
N009010 |
Cooling water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359365
|
Brady™
N009011 |
Cooling water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329385
|
Brady™
N009081 |
Demineralised Water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339385
|
Brady™
N009082 |
Demineralised Water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349385
|
Brady™
N009083 |
Demineralised Water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319445
|
Brady™
N009306 |
Drinking water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329445
|
Brady™
N009307 |
Drinking water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339445
|
Brady™
N009308 |
Drinking water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379285
|
Brady™
N008624 |
Heating return | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369285
|
Brady™
N008623 |
Heating return | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359285
|
Brady™
N008622 |
Heating return | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309285
|
Brady™
N008614 |
Heating supply | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399275
|
Brady™
N008613 |
Heating supply | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319285
|
Brady™
N008615 |
Heating supply | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329335
|
Brady™
N008838 |
Hot water return | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339335
|
Brady™
N008839 |
Hot water return | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349335
|
Brady™
N008840 |
Hot water return | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369325
|
Brady™
N008802 |
Hot water supply | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379325
|
Brady™
N008803 |
Hot water supply | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389325
|
Brady™
N008804 |
Hot water supply | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349325
|
Brady™
N008740 |
Hot water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339325
|
Brady™
N008739 |
Hot water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359325
|
Brady™
N008741 |
Hot water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC | |
Silicone | |
Roll | |
Pipe Marker | |
English | |
30 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m |
Permanent Acrylic, Permanent, Pressure-sensitive | |
White on Green | |
Polyester | |
Pipe and valve marking | |
30 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
36.09 yd. |