missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, with Pictograms, Gas

European Standard Pipemarkers reinforce safety and aid maintenance.

Brand:  Brady™ N007551

Code : NEW

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 15359105

  • £14.55 / Pack of 3
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Permanent Acrylic, Permanent, Pressure-sensitive
Warning
Card
Pipe Marker
Pipe and valve marking
English
3 Pk.
-40°C to +110°C
0.099 mm
13.98 in.
1.46 in.
16.5 mm
Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Cooling gas
Black on Ochre
Gloss
Polyester
Cooling gas
3
3 Markers/Card
-40°F to +230°F
0.0039 in.
37 x 335 mm
355 mm
37 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers