missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads -- Steam
European Standard Pipemarkers reinforce safety and aid maintenance.
£14.55 - £154.00
Specifications
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
---|---|
Color | Black on Silver |
Format | Card |
Material | Polyester |
Product Type | Pipe Marker |
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Quantity | Price | Quantity & Availability | ||
15389495
|
Brady™
N009513 |
High Pressure Steam | 26 mm | 250 mm | 30 Pk. |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15399495
|
Brady™
N009514 |
High Pressure Steam | 37 mm | 355 mm | 30 Pk. |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15309505
|
Brady™
N009515 |
High Pressure Steam | 52 mm | 450 mm | 30 Pk. |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15369495
|
Brady™
N009511 |
High Pressure Steam | 37 mm | 355 mm | 3 Pk. |
£14.55
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15329505
|
Brady™
N009520 |
HP Steam | 37 mm | 355 mm | 3 Pk. |
£14.55
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15339495
|
Brady™
N009496 |
Low Pressure Steam | 37 mm | 355 mm | 30 Pk. |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15329495
|
Brady™
N009495 |
Low Pressure Steam | 26 mm | 250 mm | 30 Pk. |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15349495
|
Brady™
N009497 |
Low Pressure Steam | 52 mm | 450 mm | 30 Pk. |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15389505
|
Brady™
N009547 |
Overheated Steam | 37 mm | 355 mm | 3 Pk. |
£14.55
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15359505
|
Brady™
N009538 |
Saturated Steam | 37 mm | 355 mm | 3 Pk. |
£14.55
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15369485
|
Brady™
N009450 |
Steam | 26 mm | 250 mm | 30 Pk. |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15379485
|
Brady™
N009451 |
Steam | 37 mm | 355 mm | 30 Pk. |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15309495
|
Brady™
N009493 |
Low Pressure Steam | 37 mm | 355 mm | 3 Pk. |
£14.55
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15349485
|
Brady™
N009448 |
Steam | 37 mm | 355 mm | 3 Pk. |
£14.55
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15389485
|
Brady™
N009452 |
Steam | 52 mm | 450 mm | 30 Pk. |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
Permanent Acrylic, Permanent, Pressure-sensitive | |
Card | |
Pipe Marker | |
English | |
33 m |
Black on Silver | |
Polyester | |
Pipe and valve marking | |
36.09 yd. |