missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms -- Air
European Standard Pipemarkers reinforce safety and aid maintenance.
£14.55
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
---|---|
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Color | White on Blue |
Finish | Gloss |
Format | Card |
Product Code | Brand | Display Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Price | Quantity & Availability | |||||
15339205
|
Brady™
N008296 |
Air |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15359215
|
Brady™
N008332 |
Compressed air |
£14.55
Pack of 3 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15309235
|
Brady™
N008449 |
Breathing air |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15399205
|
Brady™
N008323 |
Hot air |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15349225
|
Brady™
N008404 |
Instrument air |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15399235
|
Brady™
N008503 |
Return |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15319225
|
Brady™
N008368 |
Sterile compressed air |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15369235
|
Brady™
N008467 |
Sterile air |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15329245
|
Brady™
N008521 |
Vacuum |
£14.55
Pack of 3 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
White on Blue | |
Card | |
Pipe Marker | |
3 | |
3 Pk. | |
-40°C to +110°C | |
0.099 mm | |
13.98 in. | |
1.46 in. | |
16.5 mm |
Permanent Acrylic, Permanent, Pressure-sensitive | |
Gloss | |
Polyester | |
Pipe and valve marking | |
3 Markers/Card | |
-40°F to +230°F | |
0.0039 in. | |
37 x 335 mm | |
355 mm | |
37 mm |