missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand: Brady™ N009295
Code : NEW
Additional Details : Weight : 0.01100kg
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
Permanent Acrylic, Permanent, Pressure-sensitive | |
Gloss | |
Polyester | |
Polluted Water | |
3 | |
3 Markers/Card | |
-40°F to +230°F | |
0.0039 in. | |
37 x 335 mm | |
355 mm | |
37 mm |
Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Polluted water | |
White on Green | |
Card | |
Pipe Marker | |
Pipe and valve marking | |
English | |
3 Pk. | |
-40°C to +110°C | |
0.099 mm | |
13.98 in. | |
1.46 in. | |
16.5 mm |