Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water (I-Z)
European Standard Pipemarkers reinforce safety and aid maintenance.
£12.30 - £15.25
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
---|---|
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Color | White on Green |
Finish | Gloss |
Format | Card |
Product Code | Brand | Width (Metric) | Display Text | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Display Text | Price | Quantity & Availability | ||||
15309425
|
Brady™
N009222 |
250 mm | Industrial water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15329425
|
Brady™
N009224 |
450 mm | Industrial water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15309315
|
Brady™
N008718 |
250 mm | Low pressure water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15339315
|
Brady™
N008720 |
450 mm | Low pressure water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15369425
|
Brady™
N009249 |
250 mm | Mineral water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15389425
|
Brady™
N009251 |
450 mm | Mineral water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15319435
|
Brady™
N009269 |
450 mm | Non-drinking water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15399425
|
Brady™
N009267 |
250 mm | Non-drinking water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15349475
|
Brady™
N009402 |
250 mm | Plant water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15369475
|
Brady™
N009404 |
450 mm | Plant water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15359435
|
Brady™
N009294 |
250 mm | Polluted water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15379435
|
Brady™
N009296 |
450 mm | Polluted water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15389285
|
Brady™
N008637 |
250 mm | Primary circuit |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15349445
|
Brady™
N009312 |
250 mm | Process water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15369445
|
Brady™
N009314 |
450 mm | Process water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15359395
|
Brady™
N009123 |
250 mm | Pure water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15379395
|
Brady™
N009125 |
450 mm | Pure water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15309455
|
Brady™
N009321 |
250 mm | Purified water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15349315
|
Brady™
N008727 |
250 mm | Raw water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15369315
|
Brady™
N008729 |
450 mm | Raw water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15329455
|
Brady™
N009323 |
450 mm | Purified water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15379475
|
Brady™
N009420 |
250 mm | Return |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15399475
|
Brady™
N009422 |
450 mm | Return |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15369365
|
Brady™
N009024 |
250 mm | River water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15389365
|
Brady™
N009026 |
450 mm | River water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15399285
|
Brady™
N008646 |
250 mm | Secondary circuit |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15319295
|
Brady™
N008648 |
450 mm | Secondary circuit |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15389385
|
Brady™
N009114 |
250 mm | Soft water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15379265
|
Brady™
N008592 |
250 mm | Supply |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15309395
|
Brady™
N009116 |
450 mm | Soft water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15399265
|
Brady™
N008594 |
450 mm | Supply |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15329465
|
Brady™
N009384 |
250 mm | Treated water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15349465
|
Brady™
N009386 |
450 mm | Treated water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15389465
|
Brady™
N009393 |
250 mm | Waste water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15309475
|
Brady™
N009395 |
450 mm | Waste water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15369355
|
Brady™
N008999 |
450 mm | Well water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15349355
|
Brady™
N008997 |
250 mm | Well water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15399295
|
Brady™
N008682 |
250 mm | Water |
£15.25
Pack of 4 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15329305
|
Brady™
N008684 |
450 mm | Water |
£12.30
Pack of 2 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
White on Green | |
Card | |
Pipe Marker | |
English | |
-40°C to +110°C | |
0.099 mm |
Permanent Acrylic, Permanent, Pressure-sensitive | |
Gloss | |
Polyester | |
Pipe and valve marking | |
-40°F to +230°F | |
0.0039 in. |