Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand: Fisherbrand™ FB67755
Description
- Class E2 stainless steel calibration weight
- In compliance with OIML R111
Specifications
E2 | |
1 g | |
Cylindrical |
Stainless Steel | |
1 g |
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 > 1g
Spot an opportunity for improvement?Share a Content Correction