Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
£112.00 - £1355.00
Specifications
Classification E2
Material Stainless Steel
Shape Cylindrical
View More Specs
Products 12
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
15535189
View Documents
Fisherbrand™
FB67904
10 kg
£1355.00
1 unit
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
15535069
View Documents
Fisherbrand™
FB67784
50 g
£162.24
1 unit
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
15575069
View Documents
Fisherbrand™
FB67788
1 kg
£417.11
1 unit
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15585059
View Documents
Fisherbrand™
FB67779
1 g
£112.00
1 unit
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
15585069
View Documents
Fisherbrand™
FB67789
2 kg
£593.00
1 unit
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15595069
View Documents
Fisherbrand™
FB67790
5 kg
£1050.00
1 unit
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
15545069
View Documents
Fisherbrand™
FB67785
100 g
£168.00
1 unit
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
15555069
View Documents
Fisherbrand™
FB67786
200 g
£199.00
1 unit
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
15565069
View Documents
Fisherbrand™
FB67787
500 g
£327.00
1 unit
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
15525069
View Documents
Fisherbrand™
FB67783
20 g
£139.00
1 unit
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
15515069
View Documents
Fisherbrand™
FB67782
10 g
£131.00
1 unit
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
15505069
View Documents
Fisherbrand™
FB67781
5 g
£123.00
1 unit
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Description
Description
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
Specifications
Specifications
E2
Stainless Steel
Cylindrical
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate