Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1 with certificate
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
£66.30 - £977.32
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15585079
|
Fisherbrand™
|
500 g |
£223.66
1 unit |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
15535079
|
Fisherbrand™
|
10 g |
£80.30
1 unit |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
15575079
|
Fisherbrand™
|
200 g |
£132.36
1 unit |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15595079
|
Fisherbrand™
|
1 kg |
£274.54
1 unit |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
15545079
|
Fisherbrand™
|
20 g |
£88.01
1 unit |
In Stock
Log in to see stock. |
|||||
15555079
|
Fisherbrand™
|
50 g |
£91.64
1 unit |
In Stock
Log in to see stock. |
|||||
15555189
|
Fisherbrand™
|
10 kg |
£977.32
1 unit |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
15565079
|
Fisherbrand™
|
100 g |
£107.64
1 unit |
In Stock
Log in to see stock. |
|||||
15525079
|
Fisherbrand™
|
5 g |
£73.56
1 unit |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
15515089
|
Fisherbrand™
|
5 kg |
£559.68
1 unit |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
15515079
|
Fisherbrand™
|
2 g |
£66.30
1 unit |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
15505089
|
Fisherbrand™
|
2 kg |
£358.28
1 unit |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15505079
|
Fisherbrand™
|
1 g |
£70.97
1 unit |
In Stock
Log in to see stock. |
|||||
Description
- Class F1 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
Specifications
F1 | |
Stainless Steel | |
Cylindrical |